Learn More
Qorpak™ Clear Borosilicate Sample Vials with Caps
Highly resistant to heat shock and chemical leaching
Supplier:  Qorpak™ GLC05182
 View more versions of this product
Catalog No. 02992582
Only null left
Explore available promotions
Description
Description
Highly resistant to heat shock and chemical leaching
  • With high visibility
  • Available with caps made of different materials (Polypropylene hole, phenolic pulp/vinyl, phenolic PolyCone and phenolic rubber)
  • Available in volumes ranging from 0.5 to 15 dr. (1.85 to 60mL)
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Rubber
Clear
Borosilicate Glass
Highly resistant to heat and chemicals
Screw
Phenolic
81 mm
24-400
1.15 in.
29.25 mm
40 mL
3.18 in.
10 dr.
Screw Cap
144/Cs.
Product Suggestions
Product Suggestions