Learn More
Corning™ PYREXPLUS™ Media/Solution Bottles
Heavy-duty PYREXPLUS™ round bottles have a protective PVC coating to help prevent glass from shattering and reduces spills
Supplier: Corning™ 61626100
Description
Heavy duty bottle which can be used for storage as well as mixing and sampling. A protective PVC coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. Bottle comes complete with autoclavable, one-piece, green colored, polypropylene plug seal cap with drip-free pouring ring. Glass bead indicates full capacity line. Teal enameled graduations and marking spot. Thread size: GL 45. Neck opening: 29 mm I.D.
Order Info
Various other caps fit this bottle and are available for separate purchase: Polypropylene colored caps in a choice of orange, green, purple, red, and light gray for ease of sorting and identification; Vented membrane cap with 0.22ÎĽm PTFE hydrophobic membrane; PBT caps with silicone septa for syringe sampling or introduction of reagents.
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Source Reduction
- Certifications and Seals of Approval
- Renewable Energy
Specifications
3.3 oz., 100 mL | |
3.3 oz. | |
2.2 in. | |
4/Cs. | |
Non-sterile | |
40 to 80 mL | |
Cylindrical | |
3.9 in. | |
GL 45 | |
Screw Cap |
100 mL | |
Pyrexplus | |
Borosilicate Glass | |
4/Pk. | |
Green | |
Wide | |
100 mm | |
Polypropylene | |
56 mm |