Learn More
Corning™ PYREX™ VISTA™ Class B Volumetric Flasks with Polyethylene Snap-Cap
Capacity tolerances are twice those specified for Class A volumetric ware
Supplier:  Corning™ 7058150
 View more versions of this product
Catalog No. 07-770-202
Only null left
Explore available promotions
Description
Description
These Class B volumetric flasks have capacity tolerances are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue block letters make inscriptions easy to read. Snap caps are supplied with all sizes.
Specifications
Specifications
To Contain
LDPE
Borosilicate Glass
12/Cs.
B
Snap Cap
Blue
50 mL
Long
6/Pk.
Flat
Product Suggestions
Product Suggestions