Learn More
PYREX™ Bushing Type Reducing Glass Joint Adapters
With standard taper inner and outer joints for changing standard taper joint sizes
Supplier:  PYREX™ 88253424
Catalog No. 01-033H
Only null left
Explore available promotions
Description
Description
These PYREX™ bushing adapters are fabricated with heavy walls for mechanical strength and are further strengthened by the heavy, uniform rim at the top which affords a serviceable finger grip. These adapters have a 34/45 Standard Taper outer and 24/40 Standard Taper inner joints that are shorter than the conventional type, thus allowing more compact and convenient assemblies. Useful for attaching thermometers with Standard Taper inner joints.
Specifications
Specifications
For attaching thermometers with standard taper inner joints
59 mm
Tube Adapter
2.32 in.
Borosilicate Glass
24/40 Inner Bottom, 34/45 Outer Top
Product Suggestions
Product Suggestions