Learn More
Biotix Pure uTIP™ Bulk Non-sterile Pipette Tips

Pure uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk are packaged in re-sealable bags making storage and racking simple. RNase, DNase, Pyrogen, and PCR Inhibitor free.
Supplier: Biotix 63305429
Description
- Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
- Packaged in re-sealable bags making storage and racking simple.
- Biotix™ uTIP™ Pure resin Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.
- Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
- Achieved through patented technologies including FlexFit and Blade
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Re-sealable bag
- Tips must be racked by hand
- Environmental benefits include:
- Recyclable
- Certifications and Seals of Approval
Specifications
250 ÎĽL | |
Non-sterile | |
Autoclavable | |
Bulk, Bagged | |
1000 Pk. | |
Yellow | |
Universal Fit, Universal Fit Multichannel | |
uTip |
Non-filtered | |
Clear | |
Universal | |
Conductive, Fine Point, Finely Tapered, Flextop, Non-beveled Point | |
Pipette Tips | |
Single-use | |
Yes | |
Polypropylene |
Promotions
-
Biotix
Get 1 Extra Case of Pipette Tips with Purchase
Buy three qualifying cases of BioTix Pipette Tips and receive a fourth case of tips.Expires: 08/31/25 Redemption Only
-
Biotix
Get Extra Biotix Pipette Products with Purchase
When you buy $250 to $1,000 in Biotix liquid handling products, you’ll get extra pipette products at no additional cost.Expires: 06/30/26
New Offer See Full Promo Details
This product contains no safety hazards