missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Prostaglandin F2 alpha tris salt, 99%
$501.11 - $501.11
Chemical Identifiers
CAS | 38562-01-5 |
---|---|
Molecular Formula | C24H45NO8 |
Synonym | (5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt; PGF2α |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ67245LB0
|
Thermo Scientific Chemicals
J67245LB0 |
5 mg |
Each for $501.11
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
38562-01-5 | |
C24H45NO8 | |
(5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt; PGF2α |
Specifications
5 mg | |
-30°C to -10°C | |
C24H45NO8 | |
(5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt; PGF2α | |
Solid | |
NC(CO)(CO)CO.CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC(O)=O | |
475.62 |
White | |
38562-01-5 | |
475.61 | |
Prostaglandin F2 alpha tris salt, 99% | |
IYGXEHDCSOYNKY-RZHHZEQLSA-N | |
(5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]cyclopentyl]hept-5-enoic acid; 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
99% |
Safety and Handling
EINECSNumber : 254-002-3
Recommended Storage : Store at -20°C
RUO – Research Use Only