missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prostaglandin E2 98.0+%, TCI Americaâ„¢
$141.73 - $840.95
Chemical Identifiers
CAS | 363-24-6 |
---|---|
Molecular Formula | C20H32O5 |
Molecular Weight (g/mol) | 352.471 |
MDL Number | MFCD00077861 |
InChI Key | XEYBRNLFEZDVAW-ARSRFYASSA-N |
Synonym | prostaglandin e2, dinoprostone, pge2, prostin e2, prepidil, cervidil, propess, minprositin e2, minprostin e2, dinoproston |
PubChem CID | 5280360 |
ChEBI | CHEBI:15551 |
IUPAC Name | (Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid |
SMILES | CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O |
Chemical Identifiers
363-24-6 | |
352.471 | |
XEYBRNLFEZDVAW-ARSRFYASSA-N | |
5280360 | |
(Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acidShow More |
C20H32O5 | |
MFCD00077861 | |
prostaglandin e2, dinoprostone, pge2, prostin e2, prepidil, cervidil, propess, minprositin e2, minprostin e2, dinoprostonShow More | |
CHEBI:15551 | |
CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O |
Specifications
363-24-6 | |
White-Yellow | |
MFCD00077861 | |
prostaglandin e2, dinoprostone, pge2, prostin e2, prepidil, cervidil, propess, minprositin e2, minprostin e2, dinoproston | |
CCCCCC(C=CC1C(CC(=O)C1CC=CCCCC(=O)O)O)O | |
352.471 | |
CHEBI:15551 | |
≥98.0% (HPLC) | |
Prostaglandin E2 |
66°C | |
C20H32O5 | |
1 mg | |
XEYBRNLFEZDVAW-ARSRFYASSA-N | |
(Z)-7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]hept-5-enoic acid | |
5280360 | |
352.47 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : UK8000000
TSCA : No
Recommended Storage : Freezer (-20 °C)