missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic acid, ≥99.5% (GC), Honeywell™ Fluka™
Eluent additive for LC-MS
Supplier: Honeywell-Fluka 4991650ML
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Propionic acid | |
| -21°C | |
| 0.993g/mL at 25°C | |
| ∼141°C (1013hPa) | |
| 955°F | |
| Poly bottle | |
| CH3CH2COOH | |
| 50 mL | |
| 506071 | |
| Completely miscible in water | |
| CCC(=O)O | |
| 74.079 | |
| 1032 | |
| 74.08g/mol | |
| ≥99.5% (GC) |
| 79-09-4 | |
| Colorless | |
| Acidic | |
| 54°C (129.2°F) | |
| Unpleasant | |
| C3H6O2 | |
| MFCD00002756 | |
| UN3463 | |
| propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin | |
| XBDQKXXYIPTUBI-UHFFFAOYSA-N | |
| propanoic acid | |
| 2.55 (vs air) | |
| CHEBI:30768 | |
| 2.4mmHg (20°C) | |
| Liquid |
Chemical Identifiers
| 79-09-4 | |
| 74.079 | |
| XBDQKXXYIPTUBI-UHFFFAOYSA-N | |
| 1032 | |
| propanoic acid |
| C3H6O2 | |
| MFCD00002756 | |
| propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin | |
| CHEBI:30768 | |
| CCC(=O)O |
Safety and Handling
P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P235
DOTInformation : class 3 - Propionic acid
RTECSNumber : 201-176-3