missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propidium Iodide, 99%, For HPLC analysis, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219545883
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Propidium Iodide | |
Red | |
Flaky Powder | |
C27H34I2N4 | |
XJMOSONTPMZWPB-UHFFFAOYSA-M | |
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide | |
104981 | |
668.4 |
For HPLC analysis | |
25535-16-4 | |
0.25 g | |
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide | |
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-] | |
668.406 | |
CHEBI:51240 | |
99% |
Chemical Identifiers
25535-16-4 | |
668.406 | |
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodideShow More | |
CHEBI:51240 | |
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]Show More |
C27H34I2N4 | |
XJMOSONTPMZWPB-UHFFFAOYSA-M | |
104981 | |
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodideShow More |