missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega HaloLink⢠Protein Array Systems
Provides a convenient way to create protein array
Supplier: Promega G4491
Includes: Two Slide System contains HaloLink slides, HaloLink array gaskets for creating 50-well arrays, a HaloTag standard protein, anti-HaloTag antibody and cell-free expression system (TNT⢠T7 Quick Coupled Transcription/Translation System or TNT SP6 High-Yield Wheat Germ Protein Expression System. Six Slide System contains HaloLink slides, HaloLink array gaskets and anti-HaloTag antibody. Standard Protein and Fusion Protein are not included with Six Slide System but can be ordered separately.
Requires : TNT T7 Quick Coupled Transcription/Translation System or TNT SP6 High-Yield Wheat Germ Protein Expression System to use HaloLink Array Six Slide System
Description
- Analyze Protein:Protein, Protein:Drug or Protein:Nucleic Acid Interactions with HaloTag(R) Technology
- Cell-free expression for quick, single-tube production of proteins of interest for the protein array
- Irreversible binding of captured protein
Specifications
Standard Protein | |
Liquid | |
HaloLink⢠Protein Array System | |
7.3 |
-30°C to -10°C | |
Protein : protein interactions, protein: drug interactions, protein: nucleic acid interactions, antibody immobilization | |
Two slide system contains HaloLink slides, HaloLink array gaskets for creating 50-well arrays, a HaloTag standard Protein, anti-HaloTag Antibody and cell-free expression system (TNT⢠T7 Quick coupled transcription/translation system or T | |
30 μg |
- By use of this product, Researcher agrees to be bound by the terms of this limited use statement. If the researcher is not willing to accept the conditions of this limited use statement, and the product is unused, Promega will accept return of the unused product and provide the researcher with a full refund.
- Researchers may only use this product in their own research and may modify or link Promega HaloTag ligands to Promega or customer-supplied moieties. No other use or transfer of this product is authorized without the express, written consent of Promega including, without limitation, commercial use. With respect to any uses outside this label license, including any diagnostic, therapeutic or prophylactic uses, please contact Promega for supply and licensing information. Promega makes no representations or warranties of any kind, either expressed or implied, including for merchantability or fitness for a particular purpose with regards to this product. The terms of this agreement shall be governed under the laws of the State of Wisconsin, USA.
- U.S. Patent 7,429,472 and other patents pending.
- U.S. Patents 5,324,637 and 5,492,817, Australian Patent 660329 and other patents.
- For Research Use. Any use of the product for diagnostics requiring clearance or approval by the FDA may require a license under Mayo Clinic U.S. Patent 6,027,913.