Learn More
Promega DeadEnd™ Colorimetric TUNEL System DFS Item
A modified TUNEL Assay designed for simple, rapid detection of apoptotic cells in situ at the single-cell level. Measures nuclear DNA fragmentation.
Supplier:  Promega G7130
 View more versions of this product
Catalog No. PR-G7130
Only null left
Explore available promotions
Includes: DAB substrate and H2O2 plus plastic coverslips for easy sample handling
Description
Description
  • A Modified TUNEL Assay for Detection of Apoptotic Cells In Situ
  • Detects apoptosis in tissue sections or cell cultures
Specifications
Specifications
40 Reactions
Compenents are stored at different temperatures
Fluid
6.8
Detection of apoptotic cells in situ at the single-cell level
Product Suggestions
Product Suggestions