missing translation for 'onlineSavingsMsg'
Learn More

Promega Apo-ONEā„¢ Homogeneous Caspase-3/7 Assay Kit DFS Item

Highly sensitive, homogeneous fluorescent apoptosis assay for use in cultured cells and with purified caspase. Just add-mix-read.

$171.29 - $5,053.00

Specifications

Format Cuvets, 96- or 384-well Plates
Detection Method Fluorescence
View More Specs

Includes: Apo-ONE Caspase Substrate Z-DEVD-R110, 100X; Apo-ONE Homogeneous Caspase Buffer

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-G7790 DFS Item
View Documents
Promega
G7790
10 mL N/A
Only null left
 
PR-G7792 DFS Item
View Documents
Promega
G7792
1 mL N/A
Only null left
 
PR-G7791 DFS Item
View Documents
Promega
G7791
100 mL N/A
Only null left
 
Description

Description

  • Fast and Sensitive Measurement of Active Caspase-3 and -7
  • Fluorescent add-mix-read apoptosis assay for use in cultured cells and with purified caspase
  • Dramatically decrease time to results with highly sensitive and simple protocol
  • Flexible assay design allows for adaptable formats and throughput needs
Specifications

Specifications

Cuvets, 96- or 384-well Plates
Fluorescence
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'