missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 1505 Low-Profile Weight Lifters Back Support Brace

Low-Profile Weight Lifters Back Support with flexible soft foam core provides support when lifting.

$54.79 - $56.62

Specifications

Color Black
For Use With (Application) Material Handling, Construction, Warehousing/Distribution, Maintenance
Width (English) 6.25 in.
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-819-462
View Documents
Ergodyne™
11491
Small
Each for $54.79
Only null left
 
19-819-464
View Documents
Ergodyne™
11492
Medium
Each for $56.62
Only null left
 
19-819-465
View Documents
Ergodyne™
11493
Large
Each for $54.79
Only null left
 
19-819-467
View Documents
Ergodyne™
ERG11495
2X-Large
Each for $54.79
Only null left
 
Description

Description

  • 6.25 in. low-profile tapered design
  • Flexible soft foam core composition
  • Nylon jersey body fabric
  • Easy cam buckle closure system
  • Rubber track webbing
Specifications

Specifications

Black
Material Handling, Construction, Warehousing/Distribution, Maintenance
6.25 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'