missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetracyanoplatinate(II) trihydrate, 99.9% (metals basis), Pt 44.9% min

CAS: 14323-36-5 | C4H6K2N4O3Pt | 431.40 g/mol

$330.11 - $1516.40

Chemical Identifiers

CAS 14323-36-5
Molecular Formula C4H6K2N4O3Pt
Molecular Weight (g/mol) 431.40
MDL Number MFCD00011374
InChI Key KBALGZKVNFJSRU-UHFFFAOYSA-N
IUPAC Name dipotassium tetracyanoplatinumbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].N#C[Pt++](C#N)(C#N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1053603
View Documents
Thermo Scientific Chemicals
01053603
1 g
Each for $330.11
Only null left
 
AA1053606
View Documents
Thermo Scientific Chemicals
01053606
5 g
Each for $1,516.40
Only null left
 
Description

Description

Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.

EINECSNumber : 209-236-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only