Learn More
Potassium bromate, 99.5%, for analysis
CAS: 7758-01-2 | BrKO3 | 167.01 g/mol
Supplier: Thermo Scientific Chemicals 268392500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentSpecifications
Potassium bromate | |
p.a. | |
White | |
Crystalline Powder | |
99.5% min. (Iodometry) | |
KBrO3 | |
11,431 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
potassium;bromate | |
23673461 | |
167.01 | |
Analytical | |
Plastic bottle | |
3.2700g/mL |
350.0°C | |
7758-01-2 | |
5 to 9 (5% soln. at 20°C) | |
250 g | |
BrKO3 | |
MFCD00011359 | |
15, 7738 | |
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water | |
[O-]Br(=O)=O.[K+] | |
167.01 | |
CHEBI:38211 | |
99.5% | |
0.1% max. (105°C) | |
3.27 |
Chemical Identifiers
7758-01-2 | |
167.01 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
23673461 | |
potassium;bromate |
BrKO3 | |
MFCD00011359 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromatShow More | |
CHEBI:38211 | |
[O-]Br(=O)=O.[K+] |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause cancer.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-829-8
RTECSNumber : EF8725000
TSCA : TSCA
RUO – Research Use Only