Learn More
Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh
Formation of amide groups using Boc strategy, as a scavenger resin | CAS: 89551-24-6 | C7H9N | 107.156 g/mol
Supplier: Thermo Scientific Chemicals 04439206
Description
Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh is used in the formation of amide groups using Boc strategy. as a scavenger resin. C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Poly (styrene-divinylbenzene) | |
| Powder | |
| C7H9N | |
| benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
| 5 g | |
| C1=CC=C(C=C1)CN | |
| 107.156 | |
| CHEBI:40538 |
| Aminomethylated, 1% cross-linked | |
| 89551-24-6 | |
| MFCD00146442 | |
| Slightly soluble in water. | |
| WGQKYBSKWIADBV-UHFFFAOYSA-N | |
| phenylmethanamine | |
| 7504 | |
| 100 to 200 Mesh |
Chemical Identifiers
| 89551-24-6 | |
| 107.156 | |
| WGQKYBSKWIADBV-UHFFFAOYSA-N | |
| 7504 | |
| phenylmethanamine |
| C7H9N | |
| MFCD00146442 | |
| benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
| CHEBI:40538 | |
| C1=CC=C(C=C1)CN |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only

