missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Adipate 98.0+%, TCI America™

$52.92 - $311.81
Chemical Identifiers
CAS | 142-88-1 |
---|---|
Molecular Formula | C10H20N2O4 |
Molecular Weight (g/mol) | 232.28 |
MDL Number | MFCD00050650 |
InChI Key | BVEGEKOBSPXUJS-UHFFFAOYSA-N |
PubChem CID | 8905 |
IUPAC Name | hexanedioic acid;piperazine |
SMILES | C1CNCCN1.C(CCC(=O)O)CC(=O)O |
Chemical Identifiers
142-88-1 | |
232.28 | |
BVEGEKOBSPXUJS-UHFFFAOYSA-N | |
hexanedioic acid;piperazine |
C10H20N2O4 | |
MFCD00050650 | |
8905 | |
C1CNCCN1.C(CCC(=O)O)CC(=O)O |
Specifications
142-88-1 | |
C10H20N2O4 | |
25 g | |
C1CNCCN1.C(CCC(=O)O)CC(=O)O | |
232.28 | |
232.28 | |
Crystalline Powder |
White | |
MFCD00050650 | |
BVEGEKOBSPXUJS-UHFFFAOYSA-N | |
hexanedioic acid;piperazine | |
8905 | |
≥98.0% (T) | |
Piperazine Adipate |
Safety and Handling
RTECSNumber : MO1955000
TSCA : Yes