Learn More
United Scientificâ„¢ PinPointâ„¢ Pipette Controller Filter DFS Item
Filters for use with the PinPointâ„¢ Pipette Controller.
$99.43 - $99.43
Specifications
For Use With PinPoint Pipette Controller
Type Filter
Quantity 5/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Filter Type Price Quantity  
Catalog Number Mfr. No. Filter Type Price Quantity  
S012424 DFS Item
View Documents
United Scientificâ„¢
UNPINPNT05
0.22 μm
Pack of 5 Each for $99.43
Only null left
S012425 DFS Item
View Documents
United Scientificâ„¢
UNPINPNT04
0.45 μm
Pack of 5 Each for $99.43
Only null left
Description
Description
  • Ultra light and powerful pipette controller is designed for comfort as well as accuracy
  • Intuitive 6 level speed setting and sensitive triggers provide a wide range of aspirating/dispense speeds
  • Pipette controller is ergonomically designed for extended usage
  • Compatible with glass or plastic serological pipettes (0.1 to 100 mL)
  • LCD indicates battery charge and 6 speed levels
  • Fills a 25 mL pipette in less than 5 seconds
  • Lithium-ion battery charges fully within 3 hours and can provide up to 8 hours use
  • Meets the requirements of the applicable EU directives, UK regulations, UL, and CAN/CSA
Specifications
Specifications
PinPoint Pipette Controller
Filter
5/Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'