missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Microtubes for 1.1 mL Microtube System
Pre-racked 1.1mL-tall microtubes boxed to match a standard 96-well microplate footprint for multichannel pipetting or automated workstations.
Supplier: Thermo Scientific™ LSG15086
Description
Thermo Scientific™ 1.1mL Microtubes and Racked Systems consist of 1.1mL tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.
The microtubes resembles 1mL pipette tips except that they have round bottoms. Like a box of 1mL pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.Highlights:
- 8 x 12 format simplifies and speeds applications in which a multi-channel pipettor is used
- Format is also compatible with robotic workstations
- Thermo Scientific Microtube System can be centrifuged
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Non-sterile | |
Microtube | |
Thermo Scientific™ | |
1.1 mL |
Store at room temperature. | |
960 tubes | |
Pierce 1.1mL Microtubes |
For Research Use Only. Not for use in diagnostic procedures.