Learn More
Eisco™ Physics Electronics Systems Set, 18 Pieces
Physics Electronics Systems Set allows multiple electronic samples to be built for educational demonstrations and lab activities.
Supplier: Eisco™ ELECSYS1
Description
- Packaged in a reusable plastic case with custom insert and lid to keep contents organized
- Allows students to build various electronic samples such as a forward basis diode, half wave rectifier, and a Zener Diode
- Capable of supplementing a wide variety of physics lessons
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Electronics System Set | |
7 to 12 | |
Board for Electricity/Electronics, Si Diodes (2), Zener Diode, Red LED Diode, Green LED Diode, Infrared (IR) Diode, Photoresistor, Light Dependent Resistor, NTC Resistor, PTC Resistor, NPN Signal Transistor, PNP Signal Transistor, NPN Power Transistor, PNP Power Transistor, Silicon Controlled Resistor (SCR), TRIAC, Uni-junction Transistor (UJT), Plastic Case with Lid | |
24 Students |
Orange | |
Building Electronic Samples for Educational Purposes | |
18 Pk. |