missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Used for rapid determination of pH values
$68.50 - $103.84
Specifications
Format | Strip |
---|---|
For Use With (Application) | Routine pH testing |
Dimensions (L x W) | 100 x 11 mm |
Catalog Number | Mfr. No. | pH Range | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | pH Range | Price | Quantity | ||||||
09-876-19
|
Cytiva
2627990 |
3.8 to 5.5 |
Pack of 200 Each for $103.84
|
|
||||||
09-927-822
|
Cytiva
2612990 |
1 to 12 |
Pack of 200 Each for $87.04
|
|
||||||
09-927-832
|
Cytiva
2626990 |
1.8 to 3.8 |
Pack of 200 Each for $68.50
|
|
||||||
09-927-833
|
Cytiva
2628990 |
5.2 to 6.8 |
Pack of 200 Each for $72.20
|
|
||||||
09-876-20
|
Cytiva
2629990 |
6 to 8.1 |
Pack of 200 Each for $88.47
|
|
||||||
09-927-835
|
Cytiva
2630990 |
8 to 9.7 |
Pack of 200 Each for $68.50
|
|
||||||
09-876-21
|
Cytiva
2631990 |
9.5 to 12 |
Pack of 200 Each for $68.50
|
|
||||||
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Strip | |
Routine pH testing | |
100 x 11 mm |