missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, extra pure, boiling range 100-140°C

CAS: 64742-49-0
$253.96 - $726.40
Specifications
| Chemical Name or Material | Petroleum ether |
|---|---|
| Melting Point | -30.0°C |
| CAS | 64742-49-0,111-65-9,142-82-5,108-87-2,110-82-7,73513-42-5 |
| Density | 0.7400g/mL |
| Boiling Point | 100.0°C to 140.0°C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | |||||
|
AC233020025
|
Thermo Scientific Chemicals
233020025 |
2.5 L | Glass Bottle |
Each for $253.96
|
|
|||||
|
AC233020250
|
Thermo Scientific Chemicals
233020250 |
25 L | Metal Drum |
Each for $726.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Petroleum ether | |
| 64742-49-0,111-65-9,142-82-5,108-87-2,110-82-7,73513-42-5 | |
| 100.0°C to 140.0°C | |
| Authentic | |
| 1.4030 to 1.4080 | |
| 0.74 | |
| 0.76 mm2/s (25°C) | |
| Solubility in water: insoluble. | |
| Extra Pure |
| -30.0°C | |
| 0.7400g/mL | |
| 2°C | |
| Liquid | |
| 0.005% max. | |
| MFCD00081849 | |
| 14, 5489 | |
| Petroleum ether |
RUO – Research Use Only

