missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, for analysis, boiling range 80-110°C
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Supplier: Thermo Scientific Chemicals 278230025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
| Petroleum ether | |
| 0.7250g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.3980 to 1.4060 (20°C, 589nm) | |
| MFCD00081849 | |
| AFABGHUZZDYHJO-UHFFFAOYSA-N | |
| 2-methylpentane | |
| 7892 |
| 64742-49-0 | |
| 78°C to 113°C | |
| Authentic | |
| Liquid | |
| C6H14 | |
| isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
| CCCC(C)C | |
| 86.178 | |
| Analytical |
Chemical Identifiers
| 64742-49-0 | |
| 86.178 | |
| AFABGHUZZDYHJO-UHFFFAOYSA-N | |
| 7892 |
| C6H14 | |
| MFCD00081849 | |
| isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
| CCCC(C)C |
Safety and Handling
Danger
RUO – Research Use Only

