Learn More
Thermo Scientific™ PEEK Sleeves for Fused Silica HPLC Capillary Tubing, 0.015 in. I.D., orange DFS Item
Make capillary tubing connections with Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing.
Supplier:  Thermo Scientific™ UPF230DUP
Catalog No. 05-700-64
Only null left
Explore available promotions
Description
Description
Use Thermo Scientific™ PEEK Sleeves for Fused Silica Capillary Tubing to connect capillary tubing to ports designed to receive 1/16" O.D. tubing. Chemically inert and flexible, they are capable of withstanding high pressures.
Product Suggestions
Product Suggestions