Learn More
Nutmeg Oil, East-Indian Type, FCC, Spectrum™ Chemical
N1125, 8008-45-5
Supplier:  Spectrum Chemical Mfg Cor N1125500MLGL
 View more versions of this product
Catalog No. 18604862
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Nutmeg Oil, East-Indian Type, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
8008-45-5
Amber Glass Bottle
500 mL
1
1.474-1.488
FCC
Chemical Identifiers
8008-45-5
Product Suggestions
Product Suggestions