Learn More
Honeywell™ North™ Respirator Cartridges and Filters Edge
Edge
Cartridges for use with 5500, RU8500 and 7700 Series half masks as well as 5400, RU6500 and 7600 Series full facepieces.
Supplier:  Honeywell™ 7580P100
 View more versions of this product
Catalog No. 29-610-73
Only null left
Edge
Description
Description
  • Filter assemblies for use with facepiece alone or with gas and vapor cartridges available
Specifications
Specifications
NIOSH
Particulate Filter
1 Pair
Magenta
North™ full or half mask respirators
Product Suggestions
Product Suggestions