missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ Powder-Free Latex Finger Cots
GREENER_CHOICE

Latex construction seals out moisture and debris, while also withstanding abrasions and punctures. Ideal for protecting fingertips and reducing finger prints when a full glove is unnecessary.

$10.16 - $10.16

Specifications

Applications Industrial, Industrial
Color White
Disposable Single-use
Exterior Finish No Texture
Hand Ambidextrous
View More Specs

Alert:

This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-023279
View Documents
Honeywell™ North™
125LWR
Medium
Pack of 720 Each for $10.16
Case of 30 Pack for $291.30
Only null left
 
Description

Description

  • Latex construction seals out moisture and debris
  • 3 mil thickness is capable of withstanding punctures and abrasion
  • Uniform thickness ensures equal sensitivity in all critical areas
  • Very low extractables to reduce contamination from reactive ions
  • Finger cots are disposable to help reduce cross contamination
  • Provides an ideal level of protection when a full glove is unnecessary
Specifications

Specifications

Industrial, Industrial
Single-use
Ambidextrous
Powder Free
3 mil
Rolled
7 cm
White
No Texture
Latex
Finger Cot
Protecting Fingertips, Reducing Fingerprint Marks
2.8 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'