missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ Air-Purifying and Continuous Flow Respirator Replacement Part, Silicone Headstrap

Replacement part for North™ air-purifying or continuous flow respirators.

Supplier:  Honeywell™ 80845

Catalog No. 29-635-9172


Only null left
Explore available promotions

Description

Description

  • Replacement part
  • For use with APR and continuous flow respirators
  • Honeywell model 80845
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications

Specifications

Neoprene
Apr and continuous flow respirator
1 Ea.
Headstrap
80845
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions