missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonyl Acetate 99.0+%, TCI Americaâ„¢
$54.81 - $351.49
Chemical Identifiers
CAS | 143-13-5 |
---|---|
Molecular Formula | C11H22O2 |
Molecular Weight (g/mol) | 186.295 |
MDL Number | MFCD00027340 |
InChI Key | GJQIMXVRFNLMTB-UHFFFAOYSA-N |
Synonym | acetic acid, nonyl ester, n-nonyl acetate, n-nonanyl acetate, acetic acid n-nonyl ester, nonanol acetate, pelargonyl acetate, n-nonyl ethanoate, acetate c-9, nonyl ethanoate, 1-acetoxynonaneShow More |
PubChem CID | 8918 |
ChEBI | CHEBI:87511 |
IUPAC Name | nonyl acetate |
SMILES | CCCCCCCCCOC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
A004125ML
|
TCI America
A004125ML |
25 mL |
Each for $54.81
|
|
||||||
A0041500ML
|
TCI America
A0041500ML |
500 mL |
Each for $351.49
|
|
||||||
Chemical Identifiers
143-13-5 | |
186.295 | |
GJQIMXVRFNLMTB-UHFFFAOYSA-N | |
8918 | |
nonyl acetate |
C11H22O2 | |
MFCD00027340 | |
acetic acid, nonyl ester, n-nonyl acetate, n-nonanyl acetate, acetic acid n-nonyl ester, nonanol acetate, pelargonyl acetate, n-nonyl ethanoate, acetate c-9, nonyl ethanoate, 1-acetoxynonaneShow More | |
CHEBI:87511 | |
CCCCCCCCCOC(=O)C |
Specifications
143-13-5 | |
228°C | |
MFCD00027340 | |
acetic acid, nonyl ester, n-nonyl acetate, n-nonanyl acetate, acetic acid n-nonyl ester, nonanol acetate, pelargonyl acetate, n-nonyl ethanoate, acetate c-9, nonyl ethanoate, 1-acetoxynonane | |
CCCCCCCCCOC(=O)C | |
186.295 | |
CHEBI:87511 | |
≥99.0% (GC) | |
Nonyl Acetate |
Colorless | |
C11H22O2 | |
25 mL | |
GJQIMXVRFNLMTB-UHFFFAOYSA-N | |
nonyl acetate | |
8918 | |
186.30 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0735
RTECSNumber : AJ1382500
TSCA : Yes