missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, 2.0N Standardized Solution

CAS: 7697-37-2 | HNO3 | 63.01 g/mol

Supplier:  Thermo Scientific Chemicals 044528AP

 View more versions of this product

Catalog No. AA44528AP


Only null left
Explore available promotions

Description

Description

Nitric acid, 2.0N Standardized Solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H332

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only