Learn More
Nickel(II) acetate tetrahydrate, 99+%, for analysis
CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
Supplier: Thermo Scientific Chemicals 223141000
Alert:
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recycled Content
- Renewable Energy
Specifications
Nickel(II) acetate tetrahydrate | |
6018-89-9 | |
Crystalline Powder, Crystals or Flakes | |
99% min. (Complexometry) | |
(CH3CO2)2Ni·4H2O | |
14,288; 16,308 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
nickel(2+);diacetate;tetrahydrate | |
62601 | |
≥99% | |
Authentic |
250.0°C | |
White | |
100 g | |
C4H14NiO8 | |
MFCD00066973 | |
14,6498 | |
Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O | |
248.84 | |
248.86 | |
Analytical | |
Plastic bottle |
Chemical Identifiers
6018-89-9 | |
248.84 | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
62601 |
C4H14NiO8 | |
MFCD00066973 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
GHS Signal Word: Danger
RUO – Research Use Only