Learn More
Thermo Scientific™ NanoDrop™ Eight SciVault Compliance Software
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ SciVault Software for use with the NanoDrop™ Eight Microvolume Spectrophotometer. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account access control, electronic signatures, audit trails, automated data exports, and security policies.
Supplier:  Thermo Scientific™ 840371100
Catalog No. 13400533
Only null left
Explore available promotions
Description
Description
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ SciVault Software for use with the NanoDrop™ Eight Microvolume Spectrophotometer. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account access control, electronic signatures, audit trails, automated data exports, and security policies. SciVault software integrates directly into the NanoDrop Eight Software for streamlined operation.
Thermo Scientific™ SciVault™ Software helps your lab stay compliant with US FDA data regulations every step of the way.
  • Streamline management –SciVault Software integrates into NanoDrop Eight Software user interface
  • Comply with US FDA 21 CFR Part 11 Regulation – Control user account access, apply digital signatures, and review audit logs
  • Network multiple instruments – Install SciVault Software on a central computer to control privileges and view audit logs across multiple NanoDrop Eight Instruments in different labs
  • Validate your systems – IQ/OQ documents and services available to confirm your instrument is running within manufacturer’s specifications
Greener Choice Claims
  • Environmental benefits include:
    • Certifications and Seals of Approval
Specifications
Specifications
SciVault compliance software for NanoDrop Eight Spectophotometers
NanoDrop Eight Spectrophotometers
Software is designed to integrate into the NanoDrop Eight PC software user interface. Software enable users to run the NanoDrop Eight instrument in a lab that needs to comply with US FDA 21 CFR Part 11. Includes: USB flash drive containing the SciVault software for the NanoDrop Eight Spectrophotometer
Product Suggestions
Product Suggestions