Learn More
Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors
GREENER_CHOICE
Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.
$165.20 - $277.23
Specifications
Autoclavable Autoclavable
Material Polypropylene
Product Type Tubing Connector
View More Specs
Products 6
Catalog Number Mfr. No. For Use With Price Quantity  
Catalog Number Mfr. No. For Use With Price Quantity  
15-320-10A
View Documents
Thermo Scientific™
61520125
1/8 in. I.D.
Pack of 12 Each for $169.57
Case of 6 Pack for $1,017.42
Only null left
15-320-10B
View Documents
Thermo Scientific™
61520187
3/16 in. I.D.
Pack of 12 Each for $169.57
Case of 6 Pack for $1,017.42
Only null left
15-320-10C
View Documents
Thermo Scientific™
61520250
1/4 in. I.D.
Pack of 12 Each for $165.20
Case of 6 Pack for $991.20
Only null left
15-320-10D
View Documents
Thermo Scientific™
61520312
5/16 in. I.D.
Pack of 12 Each for $188.10
Case of 6 Pack for $1,128.60
Only null left
15-320-10E
View Documents
Thermo Scientific™
61520375
3/8 in.
Pack of 12 Each for $237.17
Case of 4 Pack for $948.68
Only null left
15-320-10F
View Documents
Thermo Scientific™
61520500
1/2 in. Tubing
Pack of 12 Each for $277.23
Case of 4 Pack for $1,108.92
Only null left
Description
Description
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Polypropylene construction
  • Autoclavable
Warranty: 90 days
Specifications
Specifications
Autoclavable
Polypropylene
Tubing Connector
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.