Learn More
Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps
GREENER_CHOICE
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents.
Supplier:  Thermo Scientific™ 63200010
Catalog No. 10-309
Only null left
Explore available promotions
Description
Description
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents. Double-action hinges provide extra leverage and long life. These forceps feature a scissor-type handle with a ratchet to lock tips shut and a handy clamp for chromatography columns.
  • Autoclavable
  • Feature jaws with interlocking teeth to grip objects tightly
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Straight, Serrated
Polypropylene
Forceps
Scissor Type
12
Forceps
12/Pk.
Product Suggestions
Product Suggestions