Learn More
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks

Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Supplier: Thermo Scientific™ 41020250
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
- Environmental benefits include:
- Renewable Energy
Specifications
8 oz. | |
250 mL | |
12 | |
Autoclavable | |
No | |
Non-sterile | |
Stopper No. 6 |
Flask Erlenmeyer PP 250mL | |
PPCO | |
6 | |
Reusable | |
Narrow | |
Plain |
Promotions
-
Thermo Fisher Scientific
Get 1 Extra Nalgene Product When You Buy 2
Buy two identical qualifying Thermo Scientific Nalgene products and get one extra of the same product.Expires: 06/30/26 Redemption Only