Learn More
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles

Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.
$176.03 - $176.03
Specifications
| Capacity (Metric) | 500 mL |
|---|---|
| Capacity (English) | 16 oz. |
| Color | Natural |
| Description | With stem and draw tube |
| Material | LDPE |
| Catalog Number | Mfr. No. | Closure Color | Price | Quantity | ||||||
|---|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Closure Color | Price | Quantity | ||||||
|
03-409-17F
|
Thermo Scientific™
24222500 |
Yellow |
N/A
|
|
||||||
|
03-409-17E
|
Thermo Scientific™
24223500 |
Blue |
N/A
|
|
||||||
|
03-409-17C
|
Thermo Scientific™
24225500 |
Red |
N/A
|
|
||||||
|
03-409-17D
|
Thermo Scientific™
24224500 |
Green |
N/A
|
|
||||||
Description
High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Color-coded top allows you to coordinate reagents with industry storage codes.
- Tip can be cut back to provide increased flow.
- Tight fit of draw tube and closure prevents separation when squeezing bottle.
- Flexible LDPE bottle provides soft, ergonomic squeeze.
- Sputter-free, precision stream.
- Bottle and Carton are readily recyclable in most communities.
- Manufactured from non-toxic materials.
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
| 500 mL | |
| Natural | |
| LDPE | |
| 6 | |
| No | |
| Cylindrical | |
| Color-coded Wash Bottle | |
| Polypropylene | |
| 72.9 mm | |
| Yes |
| 16 oz. | |
| With stem and draw tube | |
| 24 | |
| Yes | |
| Narrow | |
| 224 mm | |
| 8.8 in. | |
| 28 mm | |
| Screw Cap | |
| Not Autoclavable |




