Learn More
N,N'-Di-Boc-1H-pyrazole-1-carboxamidine, 98+%
CAS: 152120-54-2 | C14H22N4O4 | 310.354 g/mol
Supplier: Thermo Scientific Chemicals H5456514
Description
N,N'-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.
Solubility
Slightly soluble in water. Soluble in methanol, and chloroform.
Notes
Store at 4°C.
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Ozone-Safe or Safe for the Ozone Layer
- Renewable Energy
- Non-Toxic
Specifications
N,N'-Di-Boc-1H-pyrazole-1-carboxamidine | |
86°C to 90°C | |
MFCD01075122 | |
n,n'-di-boc-1h-pyrazole-1-carboxamidine, bis-boc-pyrazolocarboxamidine, pyrazol boc 2, n,n-bis-boc-1-guanylpyrazole, n,n'-bis-boc-1-guanylpyrazol, n,n'-bis-boc-1-guanylpyrazole, n,n'-bisboc-pyrazole-1-carboximidamide, tert-butyl tert-butoxycarbonyl amino 1h-pyrazol-1-yl methylene carbamate, tert-butyl boc-amino 1h-pyrazol-1-yl methylene carbamate, 1-n,n'-di-boc amidino pyrazole | |
QFNFDHNZVTWZED-UHFFFAOYSA-N | |
tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamate | |
6383521 | |
≥98% |
152120-54-2 | |
C14H22N4O4 | |
25 g | |
Slightly soluble in water. Soluble in methanol,and chloroform. | |
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1 | |
310.354 | |
310.35 |
Chemical Identifiers
152120-54-2 | |
310.354 | |
QFNFDHNZVTWZED-UHFFFAOYSA-N | |
6383521 | |
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1 |
C14H22N4O4 | |
MFCD01075122 | |
n,n'-di-boc-1h-pyrazole-1-carboxamidine, bis-boc-pyrazolocarboxamidine, pyrazol boc 2, n,n-bis-boc-1-guanylpyrazole, n,n'-bis-boc-1-guanylpyrazol, n,n'-bis-boc-1-guanylpyrazole, n,n'-bisboc-pyrazole-1-carboximidamide, tert-butyl tert-butoxycarbonyl amino 1h-pyrazol-1-yl methylene carbamate, tert-butyl boc-amino 1h-pyrazol-1-yl methylene carbamate, 1-n,n'-di-boc amidino pyrazoleShow More | |
tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamateShow More |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only