missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecyl mercaptan, 90%, technical
CAS: 2917-26-2 | C16H34S | 258.5 g/mol
Supplier: Thermo Scientific Chemicals 120520100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Hexadecyl mercaptan | |
18°C to 20°C | |
0.8400g/mL | |
101°C | |
88% min. (GC) | |
C16H34S | |
CH3(CH2)15SH | |
10 mL | |
0.84 | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethyl alcohol,carbon tetrachl,soluble in ether | |
CCCCCCCCCCCCCCCCS | |
258.5 | |
258.5 | |
Technical |
2917-26-2 | |
Colorless or White to Yellow | |
184°C to 191°C (7.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4610 to 1.463 | |
MFCD00011677 | |
01, 430 | |
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan | |
ORTRWBYBJVGVQC-UHFFFAOYSA-N | |
hexadecane-1-thiol | |
18015 | |
90% | |
Liquid or Waxy Solid |
Chemical Identifiers
2917-26-2 | |
258.5 | |
ORTRWBYBJVGVQC-UHFFFAOYSA-N | |
18015 | |
CCCCCCCCCCCCCCCCS |
C16H34S | |
MFCD00011677 | |
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptanShow More | |
hexadecane-1-thiol |
Safety and Handling
EINECSNumber : 220-846-6
RUO – Research Use Only