missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecyl mercaptan, 90%, technical, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 120521000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
n-Hexadecyl mercaptan | |
0.8400g/mL | |
101°C | |
88% min. (GC) | |
C16H34S | |
CH3(CH2)15SH | |
100mL | |
0.84 | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethyl alcohol,carbon tetrachl,soluble in ether | |
CCCCCCCCCCCCCCCCS | |
258.5 | |
258.5 | |
Technical |
2917-26-2 | |
184°C to 191°C (7.0mmHg) | |
Authentic | |
Glass bottle | |
1.4610 to 1.463 | |
MFCD00011677 | |
01, 430 | |
1-hexadecanethiol, hexadecanethiol, n-hexadecanethiol, cetyl mercaptan, n-hexadecyl mercaptan, hexadecyl mercaptan, unii-qr98qio1ql, 1-cetanethiol, qr98qio1ql, cetylmercaptan | |
ORTRWBYBJVGVQC-UHFFFAOYSA-N | |
hexadecane-1-thiol | |
18015 | |
90% |
Safety and Handling
EINECSNumber : 220-846-6
RUO â Research Use Only