Learn More
n-Hexadecane, 99%, pure
n-Hexadecane, 99%, C16H34, CAS Number-544-76-3, n-cetane, n-hexadecane, hexadecan, hexadekan, cetan, cetane, ccris 5833, zetan, pentadecane, methyl, unii-f8z00shp6q, 2.5L, 98.5% min. (GC), 01,172, 287.0 deg.C, 100.0, CHEBI:45296, Colorless, 0.7730g/mL, 208-878-9, 135 deg.C, 226.44, Pure | CAS: 544-76-3 | C16H34 | 226.44 g/mol
Supplier: Thermo Scientific Chemicals 120460100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Hexadecane | |
98.5 | |
18.0°C | |
0.7730g/mL | |
135°C | |
98.5% min. (GC) | |
C16H34 | |
CH3(CH2)14CH3 | |
10 L | |
0.773 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone and ether | |
CCCCCCCCCCCCCCCC | |
226.44 | |
3.032 mPa.s (25°C) | |
226.44 | |
Pure |
544-76-3 | |
100.0 | |
Colorless | |
287.0°C | |
Authentic | |
Metal Drum | |
1.4330 to 1.4350 | |
MFCD00008998 | |
01,172 | |
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl | |
DCAYPVUWAIABOU-UHFFFAOYSA-N | |
hexadecane | |
11006 | |
CHEBI:45296 | |
99% | |
Liquid |
Chemical Identifiers
544-76-3 | |
226.44 | |
DCAYPVUWAIABOU-UHFFFAOYSA-N | |
11006 | |
hexadecane |
C16H34 | |
MFCD00008998 | |
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methylShow More | |
CHEBI:45296 | |
CCCCCCCCCCCCCCCC |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-878-9
RUO – Research Use Only