Learn More
n-Heptadecane, 99%
CAS: 629-78-7 | C17H36 | 240.47 g/mol
Supplier: Thermo Scientific Chemicals 156241000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
n-Heptadecane | |
22.0°C to 24.0°C | |
0.7780g/mL | |
148°C | |
98.5% min. (GC) | |
C17H36 | |
CH3(CH2)15CH3 | |
01,173 | |
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078 | |
NDJKXXJCMXVBJW-UHFFFAOYSA-N | |
heptadecane | |
12398 | |
240.47 | |
Liquid After Melting |
629-78-7 | |
Colorless | |
302.0°C | |
Authentic | |
Glass bottle | |
1.4350 to 1.4370 | |
100 mL | |
0.778 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
CCCCCCCCCCCCCCCCC | |
240.47 | |
CHEBI:16148 | |
99% |
Chemical Identifiers
629-78-7 | |
240.47 | |
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078 | |
CHEBI:16148 | |
CCCCCCCCCCCCCCCCC |
C17H36 | |
NDJKXXJCMXVBJW-UHFFFAOYSA-N | |
12398 | |
heptadecane |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 211-108-4
RUO – Research Use Only