Learn More
N-Formylpiperidine, 99%
CAS: 2591-86-8 | C6H11NO | 113.16 g/mol
Supplier: Thermo Scientific Chemicals 119730010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Formylpiperidine | |
2591-86-8 | |
100.0 | |
Colorless to Yellow | |
222°C | |
Authentic | |
Glass bottle | |
1.4830 to 1.485 | |
1 L | |
11,244 | |
n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg | |
FEWLNYSYJNLUOO-UHFFFAOYSA-N | |
piperidine-1-carbaldehyde | |
17429 | |
113.16 | |
Liquid |
99% | |
98.5 | |
-31°C | |
1.0190g/mL | |
91°C | |
99% | |
C6H11NO | |
MFCD00006483 | |
20, 45 | |
1.019 | |
Solubility in water: freely soluble. Other solubilities: soluble in ethanol,ether,hexane | |
C1CCN(CC1)C=O | |
113.16 | |
CHEBI:42546 | |
99% |
Chemical Identifiers
2591-86-8 | |
113.16 | |
FEWLNYSYJNLUOO-UHFFFAOYSA-N | |
17429 | |
piperidine-1-carbaldehyde |
C6H11NO | |
MFCD00006483 | |
n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czgShow More | |
CHEBI:42546 | |
C1CCN(CC1)C=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat,drink or smoke when using this product.
GHS Signal Word: Danger
EINECSNumber : 219-986-0
RTECSNumber : TN0380000
TSCA : TSCA
RUO – Research Use Only