Learn More
MSA™ Advantage™ Cartridges
Lightweight for comfort, but designed for maximum protection
$18.83 - $99.26
Specifications
Certifications/Compliance OSHA, NIOSH 42CFR, Part 84, EN 14387, AS NZS 1716, GOST, DSTU
Connection Type Bayonet
Product Type Advantage™ Respirator Cartridge
For Use With (Application) Asbestos abatement, welding, sanding, grinding, painting, public events, search, rescue
For Use With (Equipment) Advantage™ 200 ls and 420 half masks and full facepiece respirator (1000, 3000 and 4000 series)
View More Specs
Products 5
Catalog Number Mfr. No. Cartridge Type Filter Type Price Quantity  
Catalog Number Mfr. No. Cartridge Type Filter Type Price Quantity  
189993687
View Documents
MSA™
815368
Mercury Vapor Cartridge P100
Pack of 2 Each for $99.26
Only null left
189993900
View Documents
MSA™
815358
Ammonia, Methylamine Cartridge N95, R951
Pack of 2 Each for $36.81
Only null left
18-999-3978
View Documents
MSA™
815369
Low Profile Particulate Cartridge P100
Pack of 2 Each for $18.83
Only null left
19-003230
View Documents
MSA™
815366
Organic Vapor, Chlorine, Sulfur Dioxide, Chlorine Dioxide, Hydrogen chloride, Hydrogen sulfide, Ammonia, Methylamine, Formaldehyde, Hydrogen fluoride, Particulates Cartridge P100
Pack for $53.32
Only null left
29811988
View Documents
MSA™
815641
Organic Vapor Cartridge GMI-P100 Filter
Pack of 2 Pair for $62.94
Only null left
Description
Description
  • Particulate, chemical, and combination cartridges
  • Bayonet-style mounting allows easy installation—cartridges lock into place with a single twist
  • Interchangeable between Advantage 200LS, Advantage 420, Advantage 1000, Advantage 4000 and Advantage 3200 respirators
Order Info
Packaged two cartridges per pack.
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirements; EN 14387. Meets OSHA 29 CFR Part 1920.134.
Specifications
Specifications
OSHA, NIOSH 42CFR, Part 84, EN 14387, AS NZS 1716, GOST, DSTU
Advantage™ Respirator Cartridge
Advantage™ 200 ls and 420 half masks and full facepiece respirator (1000, 3000 and 4000 series)
Bayonet
Asbestos abatement, welding, sanding, grinding, painting, public events, search, rescue
2/Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'