missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Multitest Slides
For laboratory use
Manufacturer: MP Biomedicals™ 096041205
Description
- Material: fragile glass
- Numbered
- Blue coating
Specifications
Fragile Glass | |
12 | |
0.27 in. | |
Laboratory use |
Multitest Slide | |
Blue | |
7 mm |