Learn More
Invitrogen™ FocalCheck™ Microspheres, 6 μm, fluorescent green/orange/dark-red ring stains
Fluorescent green/orange/dark-red ring stains
Supplier: Invitrogen™ F14806
Description
The sharp ring stains exhibited by the 6.0μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.
Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)
Specifications
Confocal Microscope Calibration | |
Orange, Dark Red, Green | |
Suspension Beads | |
0.5 mL | |
6 μm |
Store in refrigerator 2°C to 8°C and protect from light. | |
Microsphere | |
FocalCheck | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.