Learn More
Moldex™ N95 Dura-Mesh™ Molded Particulate Respirators
N95 respirators with latex two-strap design are constructed of a high-resistant shell that holds its shape longer even in hot and humid environments.
Supplier:  Moldex™ 2200N95
 View more versions of this product
Catalog No. 18-999-1449
Only null left
Explore available promotions
Description
Description
N95 respirators with latex two-strap design are constructed of a high-resistant shell that holds its shape longer even in hot and humid environments.
  • Dura-Mesh™ shell resists collapsing in heat and humidity
  • Molded nose bridge seals easily without a metal noseband
  • Facepiece is naturally contoured for less pressure
  • Straps are securely attached so they don’t break at the seal
  • Softspun™ lining for increased comfort and durability
  • Soft foam nose cushion for added comfort
  • Available in three sizes to fit more workers and increase OSHA compliance
  • NIOSH certified to have a filter efficiency of 95% or greater for non-oil based particulates
  • Certified under 42CFR84
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2009 Section 7.11.1
  • N95 Series Applications: Grinding, torch cutting, sanding, metals, sweeping, bagging, foundries, stone quarrying, agriculture, construction, cement, ceramics manufacturing, underground mining, timber/lumber mills, polishing, buffing, lime, poultry, textiles, welding, brazing, soldering
  • N95 Plus Nuisance Levels of Ozone and Organic Vapors Applications (Fisher cat #19-003-246F only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
  • N95 Plus Nuisance Levels of Acid Gas Irritants Applications (Fisher cat #19-003-246G only): Aluminum smelting, glass etching, electroplating, fiberglass manufacturing, paper processing, chemical processing, brewing operations, fertilizer production
Compliance
Certified under NIOSH 42 CRF 84 to have a filter efficiency level of at least 95% effective against particulate aerosols free of oil, up to 10X the OSHA Permissable Exposure Limit (PEL).
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Specifications
N - Not Resistant to Oil
NIOSH 42 CFR 84, N95
Cup/Molded
N95
General Purpose Particulate Respirator
No
Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Latex
None
Two-Strap Design
ANSI/ISEA 110-2009, Section 7.11.1
White
Single-use
Dura-Mesh
Medium/Large
95%
No
2200N95
20/Pk.
No
Product Suggestions
Product Suggestions