Learn More
Beckman Coulter Canada Lp MINI POLY-Q VIAL BULK PAK 2000
Supplier: Beckman Coulter 592928
Mini POLY-Q Vials bulk pack of 2 000 vials w/ caps 6mL POLY-Q vials are made of high density linear polyethylene and have a very low permeability for the organic solvents used in scintillation cocktails
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy