Learn More
Biotium Inc Mini Cell Scrapers
Supplier: Biotium 22003
Mini Cell Scrapers for harvesting cells or cell lysates from 96-, 48- and 24-well plates.Features: Size: 0.5 cm (3/16") in width and 6 cm (2 3/8") in length. Packaged in 20 sterile packs of 10 scrapers per pack. Polyethylene, disposable & sterile.