missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Milli-Q™ 7010-7015 Purification Cartridge Kit

For optimal protection of the Reverse Osmosis membrane for use with Milli-Q™ IX 7010 and Milli-Q™ IX 7015 systems, and advanced protection against airborne contaminants for Milli-Q™ IQ tank top.
$2929.16 - $2929.16
Specifications
Capacity | 25 L, 50 L and/or 100 L |
---|---|
Flow Rate | 3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015) |
For Use With (Equipment) | Milli-Q™ IX 7003/05/10/15 Water Purification System |
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
IX70XXPKT1
|
MilliporeSigma™
IX70XXPKT1 |
IX 7010-7015 Purification Cartridge Kit |
Each for $2,929.16
|
|
||||||
IX70XXPKT1H
|
MilliporeSigma™
IX70XXPKT1H |
IX H 7010-7015 Purification Cartridge Kit |
Each for $2,929.16
|
|
||||||
Description
Water Purification Consumables Kit, compatible with the Milli-Q™ IX 7010 and Milli-Q™ IX 7010 systems and includes:
- IPAK GARD™ 10-15 Pretreatment Pack
- Activated Carbon for protection against chlorine, organics, bacteria
- Depth Filter for protection against particulates and colloids
- Tank Vent Filter
- Activated Carbon for volatile organics removal
- Soda Lime for CO2 removal
- Membrane Filter to stop particulates and bacteria
Specifications
25 L, 50 L and/or 100 L | |
3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015) | |
Milli-Q™ IX 7003/05/10/15 Water Purification System |