Learn More
MilliporeSigma™ Milli-Q™ 7003-7005 Purification Cartridge Kit

For full confidence that water is not influencing your analyses, the Milli-Q™ IX pure water system provides a reliable source of high-quality Type 2 pure water to feed your labâs needs: from general laboratory uses, such as instrument feed or buffer preparation, to your critical applications, such as microbiology culture media or staining solutions
$2704.03 - $2704.03
Specifications
Capacity | 25 L, 50 L and/or 100 L |
---|---|
Flow Rate | 3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015) |
For Use With (Equipment) | Milli-Q™ IX 7003/05/10/15 Water Purification System |
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
IX700XPKT1
|
MilliporeSigma™
IX700XPKT1 |
IX 7003-7005 Purification Cartridge Kit |
Each for $2,704.03
|
|
||||||
IX700XPKT1H
|
MilliporeSigma™
IX700XPKT1H |
IX H 7003-7005 Purification Cartridge Kit |
Each for $2,704.03
|
|
||||||
Description
Water Purification Consumables Kit, compatible with the Milli-Q™ IX 7003 and Milli-Q™ IX 7005 systems and includes:
- IPAK GARD™ H 3-5 Pretreatment Pack (hard water)
- Activated Carbon for protection against chlorine, organics, bacteria
- Polyphosphate for protection against scaling and precipitates
- Depth Filter for protection against particulates and colloids
- Tank Vent Filter
- Activated Carbon for volatile organics removal
- Soda Lime for CO2 removal
- Membrane Filter to stop particulates and bacteria
Specifications
25 L, 50 L and/or 100 L | |
3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015) | |
Milli-Q™ IX 7003/05/10/15 Water Purification System |